Drukuj

Anti-PDZD2 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA315217EA 2830 PLN
ORIGTA315217
Anti-PDZD2 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: PDZD2 (PDZ domain containing 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR