Drukuj

Anti-MAPK8IP3 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA315223EA 2830 PLN
ORIGTA315223
Anti-MAPK8IP3 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: MAPK8IP3 (mitogen-activated protein kinase 8 interacting protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR