Drukuj

Anti-MPRIP Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA315225EA 2830 PLN
ORIGTA315225
Anti-MPRIP Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: MPRIP (myosin phosphatase Rho interacting protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR