Drukuj

Anti-SEPT8 Rabbit Polyclonal Antibody

ORIGTA315226EA 1680 PLN
ORIGTA315226
Anti-SEPT8 Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: SEPT8 (septin 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR