Drukuj

Anti-ZC3H4 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA315229EA 2730 PLN
ORIGTA315229
Anti-ZC3H4 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: ZC3H4 (zinc finger CCCH-type containing 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR