Drukuj

Anti-RHOBTB2 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA315231EA 2730 PLN
ORIGTA315231
Anti-RHOBTB2 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: RHOBTB2 (Rho-related BTB domain containing 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR