Drukuj

Anti-USP24 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA315240EA 2830 PLN
ORIGTA315240
Anti-USP24 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: USP24 (ubiquitin specific peptidase 24)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR