Drukuj

Anti-ABCA6 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA315248EA 2730 PLN
ORIGTA315248
Anti-ABCA6 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: ABCA6 (ATP-binding cassette, sub-family A (ABC1), member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR