Drukuj

Anti-OLFML2B Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA315296EA 2730 PLN
ORIGTA315296
Anti-OLFML2B Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: OLFML2B (olfactomedin-like 2B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR