Drukuj

Anti-OR2K2 Rabbit Polyclonal Antibody

ORIGTA315316EA 1680 PLN
ORIGTA315316
Anti-OR2K2 Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: OR2K2 (olfactory receptor, family 2, subfamily K, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR