Drukuj

Anti-OPTC Rabbit Polyclonal Antibody

ORIGTA315319EA 1680 PLN
ORIGTA315319
Anti-OPTC Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: OPTC (opticin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR