Drukuj

Anti-ARFGAP3 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA315323EA 2730 PLN
ORIGTA315323
Anti-ARFGAP3 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: ARFGAP3 (ADP-ribosylation factor GTPase activating protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR