Drukuj

Anti-OR8G1 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA315328EA 2830 PLN
ORIGTA315328
Anti-OR8G1 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: OR8G1 (olfactory receptor, family 8, subfamily G, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR