Drukuj

Anti-MMP9 Mouse Monoclonal Antibody [clone: 2H4]

Dostawca: Abnova
ABNOH00004318-M03EA 2010 PLN
ABNOH00004318-M03
Anti-MMP9 Mouse Monoclonal Antibody [clone: 2H4]
Przeciwciała
Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone: 2H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR