Drukuj
Anti-ALDH6A1 Rabbit Polyclonal Antibody
Dostawca: Abnova
ABNOH00004329-D01PEA 1940 PLN
ABNOH00004329-D01P
Anti-ALDH6A1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: ALDH6A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human