Drukuj
Anti-MNDA Mouse Polyclonal Antibody
Dostawca: Abnova
ABNOH00004332-B01PEA 2010 PLN
ABNOH00004332-B01P
Anti-MNDA Mouse Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: MNDA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human