Drukuj

Anti-MOBP Mouse Monoclonal Antibody [clone: 4C2]

Dostawca: Abnova
ABNOH00004336-M08EA 2010 PLN
ABNOH00004336-M08
Anti-MOBP Mouse Monoclonal Antibody [clone: 4C2]
Przeciwciała
Type: Primary
Antigen: MOBP
Clonality: Monoclonal
Clone: 4C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR