Drukuj

Anti-MSX2 Mouse Monoclonal Antibody [clone: 1E8]

Dostawca: Abnova
ABNOH00004488-M10AEA 2010 PLN
ABNOH00004488-M10A
Anti-MSX2 Mouse Monoclonal Antibody [clone: 1E8]
Przeciwciała
Type: Primary
Antigen: MSX2
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR