Drukuj
Anti-MTF1 Mouse Monoclonal Antibody [clone: 2E5]
Dostawca: Abnova
ABNOH00004520-M01EA 2010 PLN
ABNOH00004520-M01
Anti-MTF1 Mouse Monoclonal Antibody [clone: 2E5]
Przeciwciała
Type: Primary
Antigen: MTF1
Clonality: Monoclonal
Clone: 2E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human