Drukuj

Anti-MTF1 Mouse Monoclonal Antibody [clone: 4A5]

Dostawca: Abnova
ABNOH00004520-M07AEA 2010 PLN
ABNOH00004520-M07A
Anti-MTF1 Mouse Monoclonal Antibody [clone: 4A5]
Przeciwciała
Type: Primary
Antigen: MTF1
Clonality: Monoclonal
Clone: 4A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR