Chcesz zalogować się jeszcze raz?
Anti-MTHFD1 Rabbit Polyclonal Antibody
Dostawca: Abnova
Type: Primary
Antigen: MTHFD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
ZAMÓW
Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 µg 100 µg ABNOH00004522-D01P
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF
SPECYFIKACJE
Antigen symbol MTHFD1
Antigen name Methylenetetrahydrofolate dehydrogenase (NADP+ Dependent) 1
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype none
Western blot Yes
Antigen synonyms 5,10-methylenetetrahydrofolate cyclohydrolase, methenyltetrahydrofolate, NADP-dependent cyclohydrolase/formyltetrahydrofolate synthetase, cytoplasmic C-1-tetrahydrofolate synthase, C1-THF synthase, MTHFD1 , MTHFD, 10-formyltetrahydrofolate synthetase, methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1, 5,10-methylenetetrahydrofolate dehydrogenase, formyltetrahydrofolate synthetase , methenyltetrahydrofolate cyclohydrolase, methylenetetrahydrofolate dehydrogenase (NADP+ dependent), MTHFC
Gene ID 4522
Cross adsorption non
Temperatura przechowywania Cool
Shipping temperature Ice
Wprowadź szczegóły
Select the article for which you are looking for Certificate
Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość