Drukuj

Anti-MTM1 Rabbit Polyclonal Antibody

Dostawca: Abnova
ABNOH00004534-D01PEA 1940 PLN
ABNOH00004534-D01P
Anti-MTM1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: MTM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR