Drukuj

Anti-MUC1 Mouse Polyclonal Antibody

Dostawca: Abnova
ABNOH00004582-A01EA 1320 PLN
ABNOH00004582-A01
Anti-MUC1 Mouse Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: MUC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR