Drukuj

Anti-MUC1 Mouse Monoclonal Antibody [clone: 1B7]

Dostawca: Abnova
ABNOH00004582-M03EA 2010 PLN
ABNOH00004582-M03
Anti-MUC1 Mouse Monoclonal Antibody [clone: 1B7]
Przeciwciała
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR