Drukuj
Anti-MUC2 Mouse Monoclonal Antibody [clone: 1H2]
Dostawca: Abnova
ABNOH00004583-M08EA 2010 PLN
ABNOH00004583-M08
Anti-MUC2 Mouse Monoclonal Antibody [clone: 1H2]
Przeciwciała
Type: Primary
Antigen: MUC2
Clonality: Monoclonal
Clone: 1H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human