Drukuj
Anti-MUC2 Mouse Monoclonal Antibody [clone: 2A9]
Dostawca: Abnova
ABNOH00004583-M15EA 2010 PLN
ABNOH00004583-M15
Anti-MUC2 Mouse Monoclonal Antibody [clone: 2A9]
Przeciwciała
Type: Primary
Antigen: MUC2
Clonality: Monoclonal
Clone: 2A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human