Drukuj
Anti-MUC4 Mouse Polyclonal Antibody
Dostawca: Abnova
ABNOH00004585-A01EA 1320 PLN
ABNOH00004585-A01
Anti-MUC4 Mouse Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: MUC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human