Drukuj
Anti-MUC7 Mouse Polyclonal Antibody
Dostawca: Abnova
ABNOH00004589-A01EA 1320 PLN
ABNOH00004589-A01
Anti-MUC7 Mouse Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: MUC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human