Drukuj

Anti-MUSK Mouse Monoclonal Antibody [clone: 4E5]

Dostawca: Abnova
ABNOH00004593-M02EA 2010 PLN
ABNOH00004593-M02
Anti-MUSK Mouse Monoclonal Antibody [clone: 4E5]
Przeciwciała
Type: Primary
Antigen: MUSK
Clonality: Monoclonal
Clone: 4E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR