Drukuj

Anti-MUT Rabbit Polyclonal Antibody

Dostawca: Abnova
ABNOH00004594-D01PEA 1940 PLN
ABNOH00004594-D01P
Anti-MUT Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: MUT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR