Anti-PSG11 Mouse Polyclonal Antibody
Catalog # ABNOH00005680-B01P
Supplier: Abnova
Supplier Article Number: H00005680-B01P
Local Article Number: ABNOH00005680-B01P
Price
2 030 zł
Log in for availability
Specifications
- Antibody type:Primary
- Antigen name:Pregnancy Specific Beta-1-glycoprotein 11
- Antigen symbol:PSG11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5680
- Antigen synonyms:PSBG-11|PSG13|PSBG-13|PSG14
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: PSG11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human