Drukuj
Anti-IFITM2 Mouse Polyclonal Antibody
Dostawca: Abnova
ABNOH00010581-B01PEA 2010 PLN
ABNOH00010581-B01P
Anti-IFITM2 Mouse Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: IFITM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human