Drukuj
Anti-PARP4 Rabbit Polyclonal Antibody
BIRBORB126232-200EA 1610 PLN
BIRBORB126232-200 BIRBORB126232-100
Anti-PARP4 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: PARP4 (poly (ADP-ribose) polymerase family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human