Drukuj

Anti-MYT1 Rabbit Polyclonal Antibody

BIRBORB126251-100EA 1120 PLN
BIRBORB126251-100 BIRBORB126251-200
Anti-MYT1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: MYT1 (myelin transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR