Drukuj

Anti-PMS1 Rabbit Polyclonal Antibody

BIRBORB126261-100EA 1120 PLN
BIRBORB126261-100 BIRBORB126261-200
Anti-PMS1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: PMS1 (PMS1 postmeiotic segregation increased 1 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR