Drukuj

Anti-PPP4R2 Rabbit Polyclonal Antibody

BIRBORB126275-100EA 1120 PLN
BIRBORB126275-100 BIRBORB126275-200
Anti-PPP4R2 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: PPP4R2 (protein phosphatase 4, regulatory subunit 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR