Drukuj

Anti-HELZ2 Rabbit Polyclonal Antibody

BIRBORB126276-200EA 1610 PLN
BIRBORB126276-200 BIRBORB126276-100
Anti-HELZ2 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: HELZ2 (helicase with zinc finger 2, transcriptional coactivator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR