Drukuj

Anti-KAPCG Rabbit Polyclonal Antibody

BIRBORB126277-200EA 1610 PLN
BIRBORB126277-200 BIRBORB126277-100
Anti-KAPCG Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: KAPCG
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR