Anti-CD22 (phospho Y807) Rabbit Monoclonal Antibody [clone: Y505]
Supplier: Abcam
New Product
Certificates
About this item
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD22 (CD22 molecule)
Clonality: monoclonal
Clone: Y505
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Specifications
- Catalog No:
- ABCAAB32040-1ML
- ABCAAB32040-100
- Antigen symbol:
- CD22
- Antigen name:
- CD22 molecule
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- Y505
- Host:
- Rabbit
- Isotype:
- IgG
- Western blot:
- Yes
- Storage temperature:
- Cool
- Shipping temperature:
- Ice
Specifications
Product Information
- Catalog #Supplier #SizePack typePkAvailabilityPrice
- Supplier Article Number: AB32040-100Local Article Number: ABCAAB32040-100Specifications:
More
Specifications
Antigen symbolCD22Environmentally PreferableAntigen nameCD22 moleculeAntibody typePrimaryClonalityMonoclonalConjugationUnconjugatedCloneY505ReactivityHostRabbitIsotypeIgGWestern blotYesStorage temperatureCoolShipping temperatureIce - Supplier Article Number: AB32040-1MLLocal Article Number: ABCAAB32040-1MLSpecifications:New Product
Specifications
Antigen symbolEnvironmentally PreferableAntigen nameAntibody typeClonalityConjugationCloneReactivityHostIsotypeWestern blotStorage temperatureShipping temperatureNew Product