Anti-ATF2 Rabbit Monoclonal Antibody [clone: E243]
: Abcam
New Product
- Catalog No:
- ABCAAB32160-1ML
- ABCAAB32160-100
- Antigen symbol:
- ATF114
- Antigen name:
- Activating transcription factor 114
- Antigen synonyms:
- ATF2
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- E243
- Host:
- Rabbit
- Isotype:
- IgG
- Western blot:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoPrecipitation:
- Yes
- Storage temperature:
- Cool
- Shipping temperature:
- Ice
- SizePack typePk
- ABCAAB32160-100AB32160-100100 µlVial100 µl1 * 100 µl3 030 zł: AB32160-100: ABCAAB32160-100
Specifications
Antigen symbolATF114Environmentally PreferableAntigen nameActivating transcription factor 114Antigen synonymsATF2Antibody typePrimaryClonalityMonoclonalConjugationUnconjugatedCloneE243ReactivityHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesImmunoPrecipitationYesStorage temperatureCoolShipping temperatureIce - : AB32160-1ML: ABCAAB32160-1MLNew Product
Specifications
Antigen symbolEnvironmentally PreferableAntigen nameAntigen synonymsAntibody typeClonalityConjugationCloneReactivityHostIsotypeWestern blotImmunoChemistryImmunoPrecipitationStorage temperatureShipping temperatureNew Product