Anti-delta 1 Catenin/CAS (phospho Y228) Rabbit Monoclonal Antibody [clone: E205]
: Abcam
New Product
- Catalog No:
- ABCAAB32403-1ML
- ABCAAB32403-100
- Antigen symbol:
- CTNND1
- Antigen name:
- catenin (cadherin-associated Protein), delta 1
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- E205
- Host:
- Rabbit
- Isotype:
- IgG
- Western blot:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoPrecipitation:
- Yes
- Storage temperature:
- Cool
- Shipping temperature:
- Ice
- SizePack typePk
- ABCAAB32403-100AB32403-100100 µlVial100 µl1 * 100 µl3 310 zł: AB32403-100: ABCAAB32403-100
Specifications
Antigen symbolCTNND1Environmentally PreferableAntigen namecatenin (cadherin-associated Protein), delta 1Antibody typePrimaryClonalityMonoclonalConjugationUnconjugatedCloneE205ReactivityHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesImmunoPrecipitationYesStorage temperatureCoolShipping temperatureIce - : AB32403-1ML: ABCAAB32403-1MLNew Product
Specifications
Antigen symbolEnvironmentally PreferableAntigen nameAntibody typeClonalityConjugationCloneReactivityHostIsotypeWestern blotImmunoChemistryImmunoPrecipitationStorage temperatureShipping temperatureNew Product