Poland
Order Entry
Anti-PHD1/prolyl hydroxylase Rabbit Monoclonal Antibody [clone: EPR2746]
Anti-PHD1/prolyl hydroxylase Rabbit Monoclonal Antibody [clone: EPR2746]
 :  Abcam
New Product
 
undefined
Anti-PHD1/prolyl hydroxylase Rabbit Monoclonal Antibody [clone: EPR2746]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCAAB113077-100
  • ABCAAB113077-1ML
  • Antigen symbol:
  • EGLN2
  • Antigen name:
  • egl-9 family hypoxia-inducible factor 2
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

 

 

  •  
     
    Size
    Pack type
    Pk
     
     
  • ABCAAB113077-100
    AB113077-100
    100 µl
    Vial
    100 µl
       
       
    1 * 100 µl
    1 * 100 µl - 3 420 zł
    3 420 zł
    1 * 100 µl
       3 420 zł

     : AB113077-100
     : ABCAAB113077-100
     
     
    Specifications
    Antigen symbol
    EGLN2
    Antigen name
    egl-9 family hypoxia-inducible factor 2
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoPrecipitation
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • ABCAAB113077-1ML
    AB113077-1ML
    1 ml
    Vial
       
       
    1 * 1 mL
    1 * 1 mL - 30 610 zł
    30 610 zł
    1 * 1 mL
       30 610 zł

     : AB113077-1ML
     : ABCAAB113077-1ML
     
    New Product
    Specifications
    Antigen symbol
    Antigen name
    Environmentally Preferable
    Antibody type
    Clonality
    Conjugation
    Reactivity
    Host
    Isotype
    Western blot
    ImmunoChemistry
    ImmunoPrecipitation
    Storage temperature
    Shipping temperature
    New Product