Poland
Order Entry
Anti-Leukotriene A4 hydrolase/LTA4H Rabbit Monoclonal Antibody [clone: EPR5713]
Anti-Leukotriene A4 hydrolase/LTA4H Rabbit Monoclonal Antibody [clone: EPR5713]
 :  Abcam
New Product
 
undefined
Anti-Leukotriene A4 hydrolase/LTA4H Rabbit Monoclonal Antibody [clone: EPR5713]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCAAB133512-100
  • ABCAAB133512-1ML
  • Antigen symbol:
  • LTA4H
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Host:
  • Rabbit
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

 

 

  •  
     
    Size
    Pack type
    Pk
     
     
  • ABCAAB133512-100
    AB133512-100
    100 µl
    Vial
    100 µl
       
       
    1 * 100 µl
    1 * 100 µl - 2 890 zł
    2 890 zł
    1 * 100 µl
       2 890 zł

     : AB133512-100
     : ABCAAB133512-100
     
     
    Specifications
    Antigen symbol
    LTA4H
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Rabbit
    Isotype
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Flow cytometry
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • ABCAAB133512-1ML
    AB133512-1ML
    1 ml
    Vial
       
       
    1 * 1 mL
    1 * 1 mL - 25 980 zł
    25 980 zł
    1 * 1 mL
       25 980 zł

     : AB133512-1ML
     : ABCAAB133512-1ML
     
    New Product
    Specifications
    Antigen symbol
    Environmentally Preferable
    Antibody type
    Clonality
    Conjugation
    Reactivity
    Host
    Isotype
    Western blot
    ImmunoChemistry
    ImmunoFluorescence
    Flow cytometry
    Storage temperature
    Shipping temperature
    New Product