Drukuj

Anti-TUBA Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: DM1A]

Dostawca: Novus Biologicals
NOVUNB100-690HEA 3390 PLN
NOVUNB100-690H
Anti-TUBA Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: DM1A]
Przeciwciała
Type: Primary
Antigen: TUBA (tubulin, alpha)
Clonality: monoclonal
Clone: DM1A
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR