Drukuj

Anti-Macrophage Scavenger Receptor I Rabbit Polyclonal Antibody (DyLight® 488)

Dostawca: Novus Biologicals
NOVUNB400-159GEA 3010 PLN
NOVUNB400-159G
Anti-Macrophage Scavenger Receptor I Rabbit Polyclonal Antibody (DyLight® 488)
Przeciwciała
Type: Primary
Antigen: Macrophage Scavenger Receptor I
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR