Drukuj

Anti-HB Mouse Monoclonal Antibody [clone: 9A5]

Dostawca: Thermo Fisher Scientific
PIEAMA514719EA 0 PLN
PIEAMA514719
Anti-HB Mouse Monoclonal Antibody [clone: 9A5]
Przeciwciała
MA5-14719 targets Hemoglobin in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14719 immunogen is human Hemoglobin. MA5-14719 detects Hemoglobin which has a predicted molecular weight of approximately 15 kDa. Product MA514719 is a smaller package size of MIH9506 (formerly sold as a Seradyn product). Hemoglobin (also spelled haemoglobin and abbreviated Hb or Hgb) is the iron-containing oxygen-transport metalloprotein in the red blood cells of vertebrates and the tissues of some invertebrates. Hemoglobin in the blood is what transports oxygen from the lungs or gills to the rest of the body (i.e. the tissues) where it releases the oxygen for cell use.

Type: Primary
Antigen: HB
Clonality: Monoclonal
Clone: 9A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR