Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-Phenytoin/Dilantin Mouse Monoclonal Antibody [clone: 1P1H2]

Dostawca: Thermo Scientific
MA5-14723 targets Phenytoin (Dilantin) in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MA5-14723 immunogen is phenytoin-KLH. Product MA514723 is a smaller package size of MIP0802 (formerly sold as a Seradyn product). Phenytoin sodium is a commonly used antiepileptic. Phenytoin (fen-i-TOE-in) acts to suppress the abnormal brain activity seen in seizure by reducing electrical conductance among brain cells by stabilizing the inactive state of voltage-gated sodium channels. Aside from seizures, it is an option in the treatment of trigeminal neuralgia in the event that carbamazepine is deemed inappropriate.

Type: Primary
Antigen: Phenytoin/Dilantin
Clonality: Monoclonal
Clone: 1P1H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chemical




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514723 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol Phenytoin/Dilantin
Antigen name Phenytoin/Dilantin
Reactivity Chemical
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 1P1H2
Host Mouse
Isotype IgG1
ELISA Yes
ImmunoPrecipitation Yes
Antigen synonyms Infatabs®, Epanutin®, Dilantin®, Kapseals®, Phenytek®
Stężenie 1 mg/ml
Immunogen Phenytoin-KLH
Purification Protein A
Form Liquid
Storage buffer 0.01M potassium phosphate, 2.5mM KCl, 0.135M NaCl, 1mg/ml BSA, 0.05% sodium azide
Temperatura przechowywania Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość