Drukuj

Anti-Caffeine Mouse Monoclonal Antibody [clone: F1-P2B1G1]

Dostawca: Thermo Fisher Scientific
PIEAMA514725EA 0 PLN
PIEAMA514725
Anti-Caffeine Mouse Monoclonal Antibody [clone: F1-P2B1G1]
Przeciwciała
MA5-14725 targets Caffeine in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MA5-14725 immunogen is caffeine-3-KLH. Product MA514725 is a smaller package size of MIC0401 (formerly sold as a Seradyn product).

Type: Primary
Antigen: Caffeine
Clonality: Monoclonal
Clone: F1-P2B1G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Chemical
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR