Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-FSH-A Mouse Monoclonal Antibody [clone: 1G6D12]

Dostawca: Thermo Scientific
MA5-14726 targets Follicle Stimulating Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14726 immunogen is human Follicle Stimulating Hormone. MA5-14726 detects Follicle Stimulating Hormone which has a predicted molecular weight of approximately 13 kDa. Product MA514726 is a smaller package size of MIF2701 (formerly sold as a Seradyn product). The pituitary glycoprotein hormone family includes follicle-stimulating hormone, luteinizing hormone, chorionic gonadotropin, and thyroid-stimulating hormone. All of these glycoproteins consist of an identical alpha subunit and a hormone-specific beta subunit. This gene encodes the beta subunit of follicle-stimulating hormone. In conjunction with luteinizing hormone, follicle-stimulating hormone induces egg and sperm production. Alternative splicing results in two transcript variants encoding the same protein.

Type: Primary
Antigen: FSH-A
Clonality: Monoclonal
Clone: 1G6D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse



Zaloguj się, aby zobaczyć swoje ceny i dostępność produktów.
  Pamiętaj mnie na tym urządzeniu
 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514726 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol FSH-A
Antigen name Follicle Stimulating Hormone alpha
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 1G6D12
Host Mouse
Isotype IgG2a
Western Blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Antigen synonyms luteinizing hormone alpha chain, lutropin alpha chain, glycoprotein hormones alpha chain, follicle-stimulating hormone alpha chain, chorionic gonadotropin, alpha polypeptide, LSH-alpha, follitropin alpha chain, chorionic gonadotrophin subunit alpha, thyroid-stimulating hormone alpha chain, thyrotropin alpha chain, follicle-stimulating hormone alpha subunit, TSH-alpha, FSH-alpha, anterior pituitary glycoprotein hormones common subunit alpha, choriogonadotropin alpha chain
Gene ID 14308, 2488
Stężenie 1 mg/ml
Immunogen Human Follicle Stimulating Hormone
Purification Protein A
Form Liquid
Storage Buffer 0.01M potassium phosphate, 2.5mM KCl, 0.135M NaCl, 1mg/ml BSA, 0.05% sodium azide
Storage temperature Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping Temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość