Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Mouse Anti-Human IgG Monoclonal Antibody [clone: F37-5]

Dostawca: Thermo Scientific
MA5-14728 targets Human IgG in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14728 immunogen is human Serum IgG. Product MA514728 is a smaller package size of MII0303 (formerly sold as a Seradyn product). Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: F37-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 ug 100 µG PIEAMA514728 Blocked
Blocked Ten produkt jest zablokowany. Prosimy, skontaktuj się z Działem Obsługi Klienta, aby uzyskać więcej informacji.
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol IgG
Antigen name Immunoglobulin G
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Secondary
Clone F37-5
Host Mouse
Isotype IgG2a
ELISA Yes
ImmunoPrecipitation Yes
Stężenie 1 mg/ml
Immunogen Human Serum IgG
Purification Ion-exchange chromatography
Form Liquid
Antibody fragment Whole Antibody
Storage buffer 0.01M potassium phosphate, 2.5mM KCl, 1mg/ml BSA, 0.135M NaCl, 0.05% sodium azide
Temperatura przechowywania Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Shipping temperature Ice
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość